CH6371379628 Chem Impex International, Inc.

Z-DL-alanine-N-hydroxysuccinimide ester

  • MDL number MFCD00189303
  • Cas Number : 73488-77-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 88-100
Synonym Z-DL-Ala-OSu
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-dl-alanine-i-n-i-hydroxysuccinimide-ester/4257?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-dl-alanine-i-n-i-hydroxysuccinimide-ester/4257?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart