CH6371379630 Chem Impex International, Inc.

Z-DL-aspartic acid beta-benzyl ester

  • MDL number MFCD00237337
  • Cas Number : 29880-21-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100 - 105 °C
Purity 99
Storage temp. Store at 0 - 8 °C
Physical Form White product
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-dl-aspartic-acid-ß-benzyl-ester/4259?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-dl-aspartic-acid-ß-benzyl-ester/4259?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart