CH6371379632 Chem Impex International, Inc.

Z-DL-leucine (oil)

  • MDL number MFCD00065129
  • Cas Number : 3588-60-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Z-DL-leucine (oil)
Purity 98
Storage temp. Store at 0-8
Physical Form Yellow clear oil
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-dl-leucine-oil/4261?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-dl-leucine-oil/4261?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart