CH6371379638 Chem Impex International, Inc.

Z-His-Met-OH

  • MDL number MFCD00238467
  • Cas Number : 2641-13-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-his-met-oh/4269?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-his-met-oh/4269?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart