CH6371379639 Chem Impex International, Inc.

Na-Z-L-histidine amide

  • MDL number MFCD00238468
  • Cas Number : 16706-41-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 194-198 �C
Synonym Z-L-His-NH2
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-z-l-histidine-amide/4270?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-z-l-histidine-amide/4270?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart