CH6371379640 Chem Impex International, Inc.

Z-L-4-trans-hydroxyproline methyl ester

  • MDL number MFCD00055851
  • Cas Number : 64187-48-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Z-L-Hyp-OMe
Purity 98
Storage temp. Store at 0-8
Physical Form Light yellowish liquid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-l-4-i-trans-i-hydroxyproline-methyl-ester/4271?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-l-4-i-trans-i-hydroxyproline-methyl-ester/4271?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart