CH6371379641 Chem Impex International, Inc.

Z-L-isoleucine dicyclohexylammonium salt

  • MDL number MFCD00043243
  • Cas Number : 26699-00-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 158-164
Synonym Z-L-Ile-OH DCHA
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-l-isoleucine-dicyclohexylammonium-salt/4272?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-l-isoleucine-dicyclohexylammonium-salt/4272?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart