CH6371379642 Chem Impex International, Inc.

Z-L-alpha-aminoadipic acid

  • MDL number MFCD00237377
  • Cas Number : 24325-14-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130-135 °C
Purity 97
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-l-alpha-aminoadipic-acid/4273?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-l-alpha-aminoadipic-acid/4273?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart