CH6371379643 Chem Impex International, Inc.

Z-L-a-aminoadipic acid d-tert-butyl ester dicyclohexylammonium salt

  • MDL number MFCD00237378
  • Cas Number : 201668-30-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Z-L-Aad(OtBu)-OH DCHA
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-l-a-aminoadipic-acid-d-tert-butyl-ester-dicyclohexylammonium-salt/4274?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-l-a-aminoadipic-acid-d-tert-butyl-ester-dicyclohexylammonium-salt/4274?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart