CH6371379644 Chem Impex International, Inc.

Z-L-leucine chloromethylketone

  • MDL number MFCD00238473
  • Cas Number : 52467-54-6
  • Molecular Weight : 297.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 34-37 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-l-leucine-chloromethylketone/4275?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-l-leucine-chloromethylketone/4275?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart