CH6371379646 Chem Impex International, Inc.

Z-Leu-Trp-OH

  • MDL number MFCD00191126
  • Cas Number : 53262-00-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144 - 146 °C
Purity 99
Storage temp. Store at 0 - 8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-leu-trp-oh/4277?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-leu-trp-oh/4277?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart