CH6371379647 Chem Impex International, Inc.

Na-Z-Ne-Boc-L-lysine methyl ester

  • MDL number MFCD08272339
  • Cas Number : 2389-60-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Z-L-Lys(Boc)-OMe
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-z-i-n-i-sup-e-sup-boc-l-lysine-methyl-ester/4278?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-z-i-n-i-sup-e-sup-boc-l-lysine-methyl-ester/4278?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart