CH6371379649 Chem Impex International, Inc.

Na-Z-L-lysine amide hydrochloride

  • MDL number MFCD00238483
  • Cas Number : 112785-42-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Z-L-Lys-NH2 HCl
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-z-l-lysine-amide-hydrochloride/4281?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-z-l-lysine-amide-hydrochloride/4281?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart