CH6371379650 Chem Impex International, Inc.

Z-(Boc-aminoethyl)-L-glycine

  • MDL number MFCD00672345
  • Cas Number : 34046-07-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Z-N-(N-beta-Boc-aminoethyl)-Gly-OH
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-boc-aminoethyl-l-glycine/4282?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-boc-aminoethyl-l-glycine/4282?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart