CH6371379651 Chem Impex International, Inc.

Z-N-methyl-L-aspartic acid b-tert-butyl ester dicyclohexylammonium salt

  • MDL number MFCD00237355
  • Cas Number : 42417-70-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Z-N-Me-L-Asp(OtBu)-OH DCHA
Purity 98
Storage temp. Store at 0-8
Physical Form White solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-i-n-i-methyl-l-aspartic-acid-beta-i-tert-i-butyl-ester-dicyclohexylammonium-salt/4283?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-i-n-i-methyl-l-aspartic-acid-beta-i-tert-i-butyl-ester-dicyclohexylammonium-salt/4283?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart