CH6371379653 Chem Impex International, Inc.

Z-N-methyl-L-glutamic acid g-tert-butyl ester

  • MDL number MFCD00237356
  • Cas Number : 42417-71-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Z-N-Me-L-Glu(OtBu)-OH
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-i-n-i-methyl-l-glutamic-acid-i-tert-i-butyl-ester/4285?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-i-n-i-methyl-l-glutamic-acid-i-tert-i-butyl-ester/4285?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart