CH6371379654 Chem Impex International, Inc.

Z-b-tert-butyl-L-alanine�A

  • MDL number MFCD02259500
  • Cas Number : 88319-45-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 156-162
Synonym Z-beta-tBu-L-Ala-OH DCHA
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-beta-i-tert-i-butyl-l-alanine-font-size-2-face-symbol-middot-font-dcha/4286?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-beta-i-tert-i-butyl-l-alanine-font-size-2-face-symbol-middot-font-dcha/4286?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart