CH6371379658 Chem Impex International, Inc.

Z-L-phenylalanine methyl ester

  • MDL number MFCD00191192
  • Cas Number : 35909-92-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Z-L-Phe-OMe
Purity 97
Storage temp. Store at 0-8
Physical Form Yellowish oil
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-l-phenylalanine-methyl-ester/4292?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-l-phenylalanine-methyl-ester/4292?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart