CH6371379659 Chem Impex International, Inc.

Z-Phe-Phe-OH

  • MDL number MFCD00191139
  • Cas Number : 13122-91-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N-Carbobenzoxy-L-phenylalanyl-L-phenylalanine
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-phe-phe-oh/4293?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-phe-phe-oh/4293?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart