CH6371379660 Chem Impex International, Inc.

Z-Phe-Trp-OH

  • MDL number MFCD00191140
  • Cas Number : 16856-28-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Z-L-phenylalanyl-L-tryptophan
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-phe-trp-oh/4294?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-phe-trp-oh/4294?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart