CH6371379662 Chem Impex International, Inc.

Z-Pro-Phe-OH

  • MDL number MFCD00037327
  • Cas Number : 17350-17-3
  • Molecular Weight : 396.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 126-130° C
Purity 99
Storage temp. Store at RT
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-pro-phe-oh/4296?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-pro-phe-oh/4296?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart