CH6371379663 Chem Impex International, Inc.

Z-O-benzyl-L-serine N-hydroxysuccinimide ester

  • MDL number MFCD00237367
  • Cas Number : 98647-23-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 81 - 86 °C
Purity 99
Storage temp. Store at 0 - 8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-i-o-i-benzyl-l-serine-n-hydroxysuccinimide-ester/4298?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-i-o-i-benzyl-l-serine-n-hydroxysuccinimide-ester/4298?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart