CH6371379664 Chem Impex International, Inc.

Z-O-4-toluenesulfonyl-L-serine methyl ester

  • MDL number MFCD00237368
  • Cas Number : 1492-52-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Z-L-Ser(Tos)-OMe
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-i-o-i-4-toluenesulfonyl-l-serine-methyl-ester/4300?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-i-o-i-4-toluenesulfonyl-l-serine-methyl-ester/4300?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart