CH6371379665 Chem Impex International, Inc.

Z-b-alanine N-hydroxysuccinimide ester

  • MDL number MFCD00238420
  • Cas Number : 53733-97-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 83-87
Synonym Z-beta-Ala-OSu
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-ß-alanine-i-n-i-hydroxysuccinimide-ester/4301?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-ß-alanine-i-n-i-hydroxysuccinimide-ester/4301?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart