CH6371379668 Chem Impex International, Inc.

Z-O-tert-butyl-L-tyrosine methyl ester

  • MDL number MFCD00237370
  • Cas Number : 5068-29-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Z-L-Tyr(tBu)-OMe
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-i-o-i-i-tert-i-butyl-l-tyrosine-methyl-ester/4307?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-i-o-i-i-tert-i-butyl-l-tyrosine-methyl-ester/4307?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart