CH6371379669 Chem Impex International, Inc.

Z-Val-Ala-OH

  • MDL number MFCD00020395
  • Cas Number : 24787-89-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Z-L-valyl-L-alanine
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-val-ala-oh/4311?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-val-ala-oh/4311?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart