CH6371379670 Chem Impex International, Inc.

Z-Val-Ile-OH

  • MDL number MFCD00027065
  • Cas Number : 41486-97-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Z-L-valyl isoleucine
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-val-ile-oh/4312?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-val-ile-oh/4312?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart