CH6371379671 Chem Impex International, Inc.

Z-Val-Phe-OH

  • MDL number MFCD00020421
  • Cas Number : 19542-51-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 173-177
Synonym N-Benzyloxycarbonyl-L-valyl-L-phenylalanine
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-val-phe-oh/4313?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-val-phe-oh/4313?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart