CH6371379672 Chem Impex International, Inc.

Z-Val-Pro-OH

  • MDL number MFCD00191171
  • Cas Number : 53331-43-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-val-pro-oh/4314?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-val-pro-oh/4314?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart