CH6371379673 Chem Impex International, Inc.

Z-Val-Trp-OH

  • MDL number MFCD00238521
  • Cas Number : 18904-53-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 196-200
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-val-trp-oh/4315?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-val-trp-oh/4315?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart