CH6371379674 Chem Impex International, Inc.

Z-O-acetyl-L-serine

  • MDL number MFCD03788037
  • Cas Number : 19645-29-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Z-L-Ser(Ac)-OH
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-i-o-i-acetyl-l-serine/4316?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-i-o-i-acetyl-l-serine/4316?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart