CH6371379676 Chem Impex International, Inc.

Na-Z-Ne-Boc-D-lysine methyl ester

  • MDL number MFCD01632083
  • Cas Number : 84559-78-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 63-70
Synonym Z-D-Lys(Boc)-OMe
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-z-i-n-i-sup-epsilon-sup-boc-d-lysine-methyl-ester/4318?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-z-i-n-i-sup-epsilon-sup-boc-d-lysine-methyl-ester/4318?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart