CH6371379677 Chem Impex International, Inc.

L-Tryptophan tert-butyl ester hydrochloride

  • MDL number MFCD00238204
  • Cas Number : 16874-09-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Trp-OtBu HCl
Purity 95
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-tryptophan-i-tert-i-butyl-ester-hydrochloride/4319?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-tryptophan-i-tert-i-butyl-ester-hydrochloride/4319?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart