CH6371379678 Chem Impex International, Inc.

D-Valine ethyl ester hydrochloride

  • MDL number MFCD00191043
  • Cas Number : 73913-64-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 102-106
Synonym D-Val-OEt HCl
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/d-valine-ethyl-ester-hydrochloride/4320?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/d-valine-ethyl-ester-hydrochloride/4320?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart