CH6371379681 Chem Impex International, Inc.

D-Tyrosine tert-butyl ester

  • MDL number MFCD00191035
  • Cas Number : 87553-74-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym D-Tyr-OtBu
Storage temp. Store at 0-8
Physical Form Crystalline
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/d-tyrosine-i-tert-i-butyl-ester/4323?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/d-tyrosine-i-tert-i-butyl-ester/4323?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart