CH6371379682 Chem Impex International, Inc.

Trityl-L-phenylalanine

  • MDL number MFCD01076154
  • Cas Number : 3226-92-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 78-82
Synonym Trt-L-Phe-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/trityl-l-phenylalanine/4324?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/trityl-l-phenylalanine/4324?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart