CH6371379683 Chem Impex International, Inc.

O-Z-L-tyrosine

  • MDL number MFCD00056724
  • Cas Number : 21106-04-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Tyr(Z)-OH
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-o-i-z-l-tyrosine/4325?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-o-i-z-l-tyrosine/4325?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart