CH6371379684 Chem Impex International, Inc.

L-Serine tert-butyl ester hydrochloride

  • MDL number MFCD00270570
  • Cas Number : 106402-41-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Ser-OtBu HCl
Purity 99
Storage temp. Store at = -15
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-serine-i-tert-i-butyl-ester-hydrochloride/4326?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-serine-i-tert-i-butyl-ester-hydrochloride/4326?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart