CH6371379686 Chem Impex International, Inc.

D-Proline ethyl ester hydrochloride

  • MDL number MFCD00191003
  • Cas Number : 131477-20-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/d-proline-ethyl-ester-hydrochloride/4328?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/d-proline-ethyl-ester-hydrochloride/4328?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart