CH6371379688 Chem Impex International, Inc.

L-Methionine tert-butyl ester hydrochloride

  • MDL number MFCD00038896
  • Cas Number : 91183-71-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 146-152
Synonym L-Met-OtBu HCl
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-methionine-i-tert-i-butyl-ester-hydrochloride/4330?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-methionine-i-tert-i-butyl-ester-hydrochloride/4330?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart