CH6371379689 Chem Impex International, Inc.

Na-Z-L-tryptophan benzyl ester

  • MDL number MFCD00191163
  • Cas Number : 69876-37-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 101-105
Synonym Z-L-Trp-OBzl
Purity 99
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-z-l-tryptophan-benzyl-ester/4331?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-z-l-tryptophan-benzyl-ester/4331?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart