CH6371379690 Chem Impex International, Inc.

L-Leucine 4-nitrophenyl ester hydrochloride

  • MDL number MFCD00270558

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point > 176 �dec.)
Synonym L-Leu-ONp HCl
Purity 98
Storage temp. Store at 0-8
Physical Form Off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-leucine-4-nitrophenyl-ester-hydrochloride/4333?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-leucine-4-nitrophenyl-ester-hydrochloride/4333?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart