CH6371379695 Chem Impex International, Inc.

Chloroacetyl-DL-serine

  • MDL number MFCD00020416

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 117-125 °C (dec.)
Purity 97
Storage temp. Store at 0-8°C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/chloroacetyl-dl-serine/4338?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/chloroacetyl-dl-serine/4338?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart