CH6371379696 Chem Impex International, Inc.

Na-Fmoc-Nd-Z-L-ornithine

  • MDL number MFCD00190893
  • Cas Number : 138775-07-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-L-Orn(Z)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-fmoc-i-n-i-sup-delta-sup-z-l-ornithine/4339?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-fmoc-i-n-i-sup-delta-sup-z-l-ornithine/4339?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart