CH6371379697 Chem Impex International, Inc.

Phthaloyl-L-alanine

  • MDL number MFCD00190990
  • Cas Number : 4192-28-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Pht-L-Ala-OH
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/phthaloyl-l-alanine/4340?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/phthaloyl-l-alanine/4340?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart