CH6371379699 Chem Impex International, Inc.

Phe-Leu-OBzl

  • MDL number MFCD01632089
  • Cas Number : 63649-15-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 157-163
Synonym L-Phenylalanyl-L-leucine benzyl ester
Purity 98
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/phe-leu-obzl/4342?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/phe-leu-obzl/4342?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart