CH6371379702 Chem Impex International, Inc.

Na-Z-Ne-trifluoroacetyl-L-lysine

  • MDL number MFCD00270524
  • Cas Number : 14905-30-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 83-103
Synonym Z-L-Lys(Tfa)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-z-i-n-i-sup-epsilon-sup-trifluoroacetyl-l-lysine/4345?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-z-i-n-i-sup-epsilon-sup-trifluoroacetyl-l-lysine/4345?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart