CH6371379706 Chem Impex International, Inc.

Boc-Asp(OBzl)-Phe-OH

  • MDL number MFCD00270492
  • Cas Number : 68763-45-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 112-119
Synonym Boc-L-aspartic acid benzyl ester phenylalanine
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-asp-obzl-phe-oh/4349?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-asp-obzl-phe-oh/4349?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart