CH6371379714 Chem Impex International, Inc.

O-Acetyl-L-tyrosine

  • MDL number MFCD01075700
  • Cas Number : 6636-22-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 197-204 �C
Synonym L-Tyr(Ac)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-o-i-acetyl-l-tyrosine/4357?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-o-i-acetyl-l-tyrosine/4357?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart