CH6371379717 Chem Impex International, Inc.

Z-L-aspartic acid dibenzyl ester

  • MDL number MFCD00056753
  • Cas Number : 5241-60-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 56-63
Synonym Z-L-Asp(OBzl)-OBzl
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-l-aspartic-acid-dibenzyl-ester/4360?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-l-aspartic-acid-dibenzyl-ester/4360?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart